siRNA Details
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110275.1 | NP_001103745.1 | 9 | 1001 | ||
NM_001110276.1 | NP_001103746.1 | 9 | 1001 | |||
NM_010587.2 | NP_034717.2 | 9 | 1001 | |||
GenBank mRNA | AF132478.1 | - | 8 | 740 | ||
AF132481.1 | - | 8 | 740 | |||
AK155397.1 | - | 9 | 956 | |||
AK160949.1 | - | 9 | 1002 | |||
BC062938.1 | - | 9 | 1001 | |||
BC066105.1 | - | 9 | 863 |

More Information
Other Species Targeted

Gene Ontology Categories:
