siRNA Details
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010593.2 | NP_034723.1 | 6 | 1132 | ||
GenBank mRNA | AK145895.1 | - | 6 | 1117 | ||
AK170934.1 | - | 6 | 1105 | |||
AK171501.1 | - | 1256 | ||||
BC040757.1 | - | 6 | 1081 | 4216772 | ||
BC094461.1 | - | 6 | 1085 | 4459882 | ||
M90365.1 | - | 4 | 597 |

More Information
Other Species Targeted

Gene Ontology Categories:
