siRNA Details
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010593.2 | NP_034723.1 | 11 | 2160 | ||
GenBank mRNA | AK145895.1 | - | 11 | 2145 | ||
AK170934.1 | - | 11 | 2133 | |||
BC040757.1 | - | 11 | 2109 | 4216772 | ||
BC094461.1 | - | 11 | 2113 | 4459882 | ||
M90365.1 | - | 9 | 1625 |
