siRNA Details
Gene Symbol: Kcnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008430.2 | NP_032456.2 | 2 | 987 | ||
GenBank mRNA | AF033017.1 | - | 2 | 919 | ||
AK137224.1 | - | 2 | 901 | |||
BC003729.1 | - | 2 | 684 | 3592756 | ||
BG088333.2 | - | 1 | 277 |
