siRNA Details
Gene Symbol: Kcnq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008434.2 | NP_032460.2 | 4 | 753 | ||
GenBank mRNA | AB079603.1 | - | 4 | 456 | ||
AK154244.1 | - | 4 | 727 | |||
AY331142.1 | - | 4 | 668 | |||
BC045142.1 | - | 4 | 753 | 6314642 | ||
BC055304.1 | - | 4 | 708 | 6397978 | ||
U70068.1 | - | 4 | 559 |
