siRNA Details
Gene Symbol: Lamb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277928.1 | NP_001264857.1 | 15 | 2202 | ||
NM_008484.2 | NP_032510.2 | 15 | 2241 | |||
GenBank mRNA | AK155769.1 | - | 3054 | |||
BC008516.1 | - | 15 | 2202 | 3592888 | ||
CX237661.1 | - | 1 | 104 | |||
U43298.1 | - | 15 | 2217 |
