siRNA Details
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286181.1 | NP_001273110.1 | 7 | 1328 | ||
NM_001286182.1 | NP_001273111.1 | 7 | 1285 | |||
NM_010728.3 | NP_034858.2 | 6 | 1705 | |||
GenBank mRNA | AK135213.1 | - | 6 | 836 | ||
AK141995.1 | - | 6 | 1591 | |||
AK159199.1 | - | 6 | 1508 | |||
AK159241.1 | - | 6 | 1484 | |||
AK159263.1 | - | 6 | 1545 | |||
AK159368.1 | - | 6 | 1467 | |||
AK159682.1 | - | 6 | 1418 | |||
AK159796.1 | - | 6 | 1479 | |||
AK160096.1 | - | 6 | 1479 | |||
AK164634.1 | - | 7 | 1286 | |||
AK166279.1 | - | 6 | 1413 | |||
AK166386.1 | - | 1549 | ||||
AK166442.1 | - | 1705 | ||||
BC018439.1 | - | 6 | 1556 | 2655752 | ||
M65142.1 | - | 6 | 1473 | |||
M65143.1 | - | 3 | 332 |
