siRNA Details
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111311.1 | NP_001104781.1 | 4 | 425 | ||
NM_001111312.1 | NP_001104782.1 | 4 | 312 | |||
NM_008515.4 | NP_032541.1 | 12 | 869 | |||
GenBank mRNA | AF045573.1 | - | 12 | 806 | ||
AK041162.1 | - | 4 | 263 | |||
AK044174.1 | - | 10 | 604 | |||
AK077637.1 | - | 4 | 430 | |||
AK133661.1 | - | 4 | 405 | |||
AK134120.1 | - | 4 | 417 | |||
BC144955.1 | - | 4 | 278 | |||
BC144956.1 | - | 14 | 827 | 9053504 | ||
BC145642.1 | - | 14 | 827 | 40130945 | ||
BY739242.1 | - | 4 | 361 |
