siRNA Details
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111311.1 | NP_001104781.1 | 7 | 853 | ||
NM_001111312.1 | NP_001104782.1 | 8 | 812 | |||
NM_008515.4 | NP_032541.1 | 18 | 1555 | |||
GenBank mRNA | AF045573.1 | - | 18 | 1492 | ||
AK041162.1 | - | 8 | 763 | |||
AK077637.1 | - | 7 | 858 | |||
AK133661.1 | - | 7 | 834 | |||
AK134120.1 | - | 7 | 845 | |||
AK162191.1 | - | 2 | 176 | |||
BC144955.1 | - | 9 | 892 | |||
BC144956.1 | - | 20 | 1513 | 9053504 | ||
BC145642.1 | - | 20 | 1513 | 40130945 |
