siRNA Details
Gene Symbol: Usp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130879.2 | NP_570949.2 | 3 | 538 | ||
GenBank mRNA | AK052297.1 | - | 3 | 479 | ||
AK133251.1 | - | 4 | 555 | |||
AK166158.1 | - | 3 | 541 | |||
AK220190.1 | - | 3 | 499 | |||
BC021769.1 | - | 3 | 497 | |||
BC145295.1 | - | 4 | 529 | |||
BY734570.1 | - | 3 | 538 |
