siRNA Details
Gene Symbol: Edaradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133643.4 | NP_598398.3 | 4 | 658 | ||
GenBank mRNA | AF358671.1 | - | 4 | 652 | ||
AK041545.1 | - | 4 | 592 | |||
AK042128.1 | - | 4 | 316 | |||
AK042414.1 | - | 4 | 621 | |||
AK075786.1 | - | 4 | 294 | |||
AY071863.1 | - | 4 | 156 | |||
BC030683.1 | - | 4 | 234 | 1344937 |
