siRNA Details
Gene Symbol: Edaradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133643.4 | NP_598398.3 | 6 | 862 | ||
GenBank mRNA | AF358671.1 | - | 6 | 856 | ||
AK041545.1 | - | 6 | 796 | |||
AK042128.1 | - | 6 | 520 | |||
AK042414.1 | - | 6 | 825 | |||
AK075786.1 | - | 6 | 498 | |||
AK148351.1 | - | 6 | 684 | |||
AY071863.1 | - | 6 | 360 | |||
BC030683.1 | - | 6 | 438 | 1344937 |
