siRNA Details
Gene Symbol: Mllt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252560.1 | NP_001239489.1 | 14 | 2000 | ||
NM_001252561.1 | NP_001239490.1 | 15 | 2051 | |||
NM_010804.4 | NP_034934.2 | 14 | 1855 | |||
GenBank mRNA | AF010135.1 | - | 14 | 1898 | ||
AK045042.1 | - | 14 | 1756 | |||
AK052207.1 | - | 5 | 786 | |||
AK154320.1 | - | 14 | 2001 | |||
AK220504.1 | - | 16 | 2107 | |||
BC055298.1 | - | 15 | 2000 | |||
BC070475.1 | - | 14 | 1821 | 30638433 |
