siRNA Details
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190817.1 | NP_001177746.1 | 13 | 4605 | ||
GenBank mRNA | BC031636.1 | - | 1 | 297 | ||
BC080300.1 | - | 13 | 4586 |
Gene Symbol: Mllt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252560.1 | NP_001239489.1 | 20 | 2816 | ||
NM_001252561.1 | NP_001239490.1 | 21 | 2867 | |||
NM_010804.4 | NP_034934.2 | 20 | 2671 | |||
GenBank mRNA | AF010135.1 | - | 20 | 2714 | ||
AK045042.1 | - | 20 | 2572 | |||
AK052207.1 | - | 11 | 1602 | |||
AK081146.1 | - | 1 | 244 | |||
AK147388.1 | - | 21 | 5226 | |||
AK154320.1 | - | 20 | 2817 | |||
AK220504.1 | - | 22 | 2923 | |||
BC055298.1 | - | 21 | 2816 | |||
BC070475.1 | - | 20 | 2637 | 30638433 |
