siRNA Details
Gene Symbol: Mut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008650.3 | NP_032676.2 | 13 | 2328 | ||
GenBank mRNA | AK012766.1 | - | 1 | 38 | ||
AK029425.1 | - | 1 | 92 | |||
AK146309.1 | - | 13 | 2288 | |||
AK148304.1 | - | 2 | 92 | |||
BC019175.1 | - | 13 | 2310 | 5007694 | ||
X51941.1 | - | 12 | 2239 |
