siRNA Details
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198587.3 | NP_001185516.2 | 5, 6 | 1896 | ||
NM_172544.3 | NP_766132.2 | 4, 5 | 786 | |||
GenBank mRNA | AK032126.1 | - | 4, 5 | 778 | ||
AK078216.1 | - | 6, 7 | 2208 | |||
AK082055.1 | - | 6, 7 | 2208 | |||
AK129205.1 | - | 3, 4 | 659 | |||
BC060719.1 | - | 6, 7 | 1471 | 6406001 | ||
CF744587.1 | - | 2, 3 | 403 |
