siRNA Details
Gene Symbol: Nsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008739.3 | NP_032765.3 | 13 | 5065 | ||
GenBank mRNA | CB247038.1 | - | 5 | 616 | ||
CD806124.1 | - | 1 | 93 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008739.3 | NP_032765.3 | 13 | 5065 | ||
GenBank mRNA | CB247038.1 | - | 5 | 616 | ||
CD806124.1 | - | 1 | 93 |