siRNA Details
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008831.4 | NP_032857.1 | 2 | 130 | ||
GenBank mRNA | AK002714.1 | - | 1 | 76 | ||
AK010619.1 | - | 2 | 73 | |||
AK150956.1 | - | 2 | 75 | |||
AK151073.1 | - | 2 | 75 | |||
AK152933.1 | - | 2 | 77 | |||
AK160490.1 | - | 2 | 69 | |||
AK168248.1 | - | 2 | 78 | |||
AK168656.1 | - | 2 | 69 | |||
AK178178.1 | - | 2 | 84 | |||
BC083354.1 | - | 1 | 52 | 5715690 | ||
BC089034.1 | - | 1 | 54 | 30675988 | ||
BY333636.1 | - | 2 | 131 | |||
X78682.1 | - | 1 | 58 |
