siRNA Details
Gene Symbol: Pmp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076005.1 | - | 2 | 175 | ||
AK171745.1 | - | 2 | 172 | |||
BC010765.1 | - | 2 | 198 | 4225316 | ||
BY092580.1 | - | 2 | 281 | |||
BY235328.1 | - | 2 | 225 | |||
BY235583.1 | - | 2 | 251 | |||
M32240.1 | - | 2 | 225 | |||
Z38110.1 | - | 1 | 39 |
