siRNA Details
Gene Symbol: Pmp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076005.1 | - | 5 | 502 | ||
AK171745.1 | - | 5 | 499 | |||
BC010765.1 | - | 5 | 525 | 4225316 | ||
M32240.1 | - | 5 | 552 | |||
Z38110.1 | - | 4 | 366 |
