siRNA Details
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011139.2 | NP_035269.2 | 2, 3 | 165 | ||
GenBank mRNA | AK153697.1 | - | 255 | |||
L14677.1 | - | 2, 3 | 163 | |||
Z18537.1 | - | 2, 3 | 166 |
