siRNA Details
Gene Symbol: Ppp2r3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163415.1 | NP_001156887.1 | 1124 | |||
NM_011154.2 | NP_035284.2 | 890 | ||||
GenBank mRNA | AF050165.1 | - | 887 | |||
AK054122.1 | - | 1263 | ||||
AK143212.1 | - | 1167 | ||||
BC027415.1 | - | 867 | 4235453 | |||
BC059852.1 | - | 1155 | 6402777 | |||
BC095938.1 | - | 45 | ||||
BC096544.1 | - | 570 | 6834351 |
