siRNA Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008917.3 | NP_032943.2 | 3 | 281 | ||
GenBank mRNA | AF071025.1 | - | 3 | 298 | ||
AF087568.1 | - | 3 | 259 | |||
AF326557.1 | - | 3 | 265 | |||
AF326558.1 | - | 3 | 265 | |||
AK014561.1 | - | 3 | 263 | |||
AK148314.1 | - | 3 | 256 | |||
AK151055.1 | - | 3 | 256 | |||
AK151107.1 | - | 3 | 267 | |||
AK152474.1 | - | 3 | 265 | |||
AK152940.1 | - | 3 | 267 | |||
AK153106.1 | - | 3 | 262 | |||
AK153316.1 | - | 3 | 262 | |||
AK169579.1 | - | 4 | 356 | |||
AK169658.1 | - | 4 | 362 | |||
AK171668.1 | - | 3 | 263 | |||
CB600397.1 | - | 3 | 284 |
