siRNA Details
Gene Symbol: Psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011184.4 | NP_035314.3 | 2 | 174 | ||
GenBank mRNA | AF055983.1 | - | 2 | 92 | ||
AF060089.1 | - | 2 | 60 | |||
AI303320.1 | - | 2 | 173 | |||
AK002877.1 | - | 2 | 105 | |||
AK165424.1 | - | 2 | 127 | |||
AK168495.1 | - | 2 | 79 | |||
AK169577.1 | - | 2 | 77 | |||
AK171669.1 | - | 2 | 83 | |||
AK172369.1 | - | 2 | 83 | |||
BC091743.1 | - | 2 | 64 | 5030475 | ||
BY159585.1 | - | 2 | 175 |
