siRNA Details
Gene Symbol: Bcas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166642.1 | NP_001160114.1 | 13 | 1075 | ||
NM_001166643.1 | NP_001160115.1 | 12 | 989 | |||
NM_138681.4 | NP_619622.3 | 13 | 1075 | |||
GenBank mRNA | AF313800.1 | - | 12 | 969 | ||
AK032423.1 | - | 13 | 1026 | |||
AK034117.1 | - | 13 | 1055 | |||
AK157222.1 | - | 13 | 1025 | |||
AK160127.1 | - | 12 | 968 | |||
AK171964.1 | - | 12 | 942 | |||
BC028339.1 | - | 12 | 963 | |||
BC038173.1 | - | 11 | 932 | |||
BC050233.1 | - | 12 | 968 | |||
BC060690.1 | - | 12 | 952 | |||
BC080775.1 | - | 11 | 936 | |||
BC086693.1 | - | 12 | 956 | |||
BC118965.2 | - | 12 | 989 | 40097104 | ||
BC118966.1 | - | 12 | 971 | 40097105 |
