siRNA Details
Gene Symbol: Ptgis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008968.3 | NP_032994.1 | 2 | 269 | ||
GenBank mRNA | AB001607.1 | - | 2 | 187 | ||
AK048059.1 | - | 322 | ||||
AK076585.1 | - | 2 | 270 | |||
BC062151.1 | - | 2 | 276 | 6512164 |
