siRNA Details
Gene Symbol: Pth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020623.2 | NP_065648.1 | 2 | 141 | ||
GenBank mRNA | AK138677.1 | - | 2 | 139 | ||
BC099456.1 | - | 2 | 126 | 30845904 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020623.2 | NP_065648.1 | 2 | 141 | ||
GenBank mRNA | AK138677.1 | - | 2 | 139 | ||
BC099456.1 | - | 2 | 126 | 30845904 |