siRNA Details
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104616.1 | NP_001098086.1 | 4 | 309 | ||
NM_001104617.1 | NP_001098087.1 | 4 | 715 | |||
NM_009041.3 | NP_033067.2 | 4 | 715 | |||
GenBank mRNA | AK016711.1 | - | 4 | 386 | ||
AK088292.1 | - | 4 | 422 | |||
AK146423.1 | - | 4 | 373 | |||
AK156370.1 | - | 4 | 401 | |||
AK162204.1 | - | 4 | 287 | |||
AK168449.1 | - | 4 | 386 | |||
BC002041.1 | - | 4 | 360 | |||
BC026786.1 | - | 4 | 388 | |||
BC034438.1 | - | 4 | 541 | |||
BC053417.1 | - | 4 | 389 | 6707873 | ||
DV662898.1 | - | 4 | 309 | |||
X60672.1 | - | 4 | 328 |
