siRNA Details
Gene Symbol: Satb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163630.1 | NP_001157102.1 | 3 | 604 | ||
NM_001163631.1 | NP_001157103.1 | 3 | 472 | |||
NM_001163632.1 | NP_001157104.1 | 3 | 445 | |||
NM_009122.2 | NP_033148.2 | 3 | 495 | |||
GenBank mRNA | AK037740.1 | - | 3 | 495 | ||
AK088563.1 | - | 3 | 471 | |||
AK134040.1 | - | 3 | 475 | |||
AK158518.1 | - | 3 | 605 | |||
BB635001.1 | - | 3 | 445 | |||
BC011132.1 | - | 3 | 355 | 4164993 | ||
CT010371.1 | - | 2 | 283 | |||
U05252.1 | - | 3 | 481 |
