siRNA Details
Gene Symbol: Sh3gl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277954.1 | NP_001264883.1 | 6 | 765 | ||
NM_001277955.1 | NP_001264884.1 | 6 | 573 | |||
NM_017400.6 | NP_059096.3 | 6 | 772 | |||
GenBank mRNA | AK012114.1 | - | 515 | |||
BC027096.1 | - | 772 | 5254139 | |||
BC100453.1 | - | 610 | 6770826 | |||
U58887.1 | - | 752 |
