siRNA Details
Gene Symbol: St8sia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011374.2 | NP_035504.2 | 5 | 1546 | ||
GenBank mRNA | AK042207.1 | - | 5 | 1402 | ||
AK046188.1 | - | 5 | 1486 | |||
AK052444.1 | - | 5 | 1197 | |||
BC024821.1 | - | 5 | 1546 | 5357151 | ||
L38677.1 | - | 5 | 1170 | |||
X84235.1 | - | 5 | 893 |
