siRNA Details
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009190.2 | NP_033216.2 | 10 | 1490 | ||
GenBank mRNA | AK034641.1 | - | 148 | |||
AK151023.1 | - | 10 | 1444 | |||
AK152131.1 | - | 10 | 1477 | |||
AK165594.1 | - | 1435 | ||||
AK168270.1 | - | 10 | 1381 | |||
BC003799.1 | - | 10 | 1397 | 3488471 | ||
U10119.1 | - | 10 | 1441 |

More Information
Other Species Targeted

Gene Ontology Categories:
