siRNA Details
Gene Symbol: Siglec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011426.3 | NP_035556.3 | 16 | 4950 | ||
GenBank mRNA | AK131121.1 | - | 2 | 526 | ||
BC141335.1 | - | 16 | 4544 | 9055947 | ||
BC141336.1 | - | 16 | 4540 | 9055948 |
