siRNA Details
Gene Symbol: Ccdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175430.4 | NP_780639.1 | 7, 8 | 2177 | ||
GenBank mRNA | AK046971.1 | - | 7, 8 | 2180 | ||
AK054172.1 | - | 8, 9 | 1691 | |||
BC072591.1 | - | 9, 10 | 1906 | |||
BC085159.1 | - | 9, 10 | 1937 | 30533905 |
