siRNA Details
Gene Symbol: Stat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132675.1 | - | 12, 13 | 1259 | ||
AK157083.1 | - | 11, 12 | 1133 | |||
BC052907.1 | - | 11, 12 | 1138 | |||
BC098499.1 | - | 11, 12 | 1126 | 30074608 | ||
U06923.1 | - | 11, 12 | 1132 | |||
U09351.1 | - | 11, 12 | 1583 |
