siRNA Details
Gene Symbol: Stat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132675.1 | - | 24, 25 | 2375 | ||
BC052907.1 | - | 23, 24 | 2255 | |||
BC098499.1 | - | 23, 24 | 2242 | 30074608 | ||
BF020635.1 | - | 1, 2 | 295 | |||
U06923.1 | - | 23, 24 | 2248 | |||
U09351.1 | - | 23, 24 | 2699 |
