siRNA Details
Gene Symbol: Gipc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148951.1 | NP_683753.1 | 6 | 829 | ||
GenBank mRNA | AB074494.1 | - | 6 | 829 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gipc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148951.1 | NP_683753.1 | 6 | 829 | ||
GenBank mRNA | AB074494.1 | - | 6 | 829 |