siRNA Details
Gene Symbol: Syt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009307.3 | NP_033333.2 | 4 | 628 | ||
GenBank mRNA | AF257304.1 | - | 3 | 383 | ||
AK036357.1 | - | 4 | 629 | |||
BC027019.1 | - | 3 | 389 | 5362352 | ||
D37793.1 | - | 2 | 262 |
