siRNA Details
Gene Symbol: Gfy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195255.1 | NP_001182184.1 | 3 | 1395 | ||
GenBank mRNA | AB845286.1 | - | 1 | 1122 | ||
BM934591.1 | - | 1 | 279 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gfy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195255.1 | NP_001182184.1 | 3 | 1395 | ||
GenBank mRNA | AB845286.1 | - | 1 | 1122 | ||
BM934591.1 | - | 1 | 279 |