siRNA Details
Gene Symbol: Pcdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081377.2 | NP_001074846.1 | 1 | 1246 | ||
NM_001271798.1 | NP_001258727.1 | 2 | 1795 | |||
NM_001271799.1 | NP_001258728.1 | 2 | 1268 | |||
NM_001271800.1 | NP_001258729.1 | 1 | 1246 | |||
GenBank mRNA | AK147451.1 | - | 2 | 1795 | ||
BC060736.1 | - | 2 | 1228 | |||
BC157952.1 | - | 1 | 1163 | 9088126 | ||
BC158114.1 | - | 1 | 1166 | 9007391 | ||
BC158117.1 | - | 1 | 1166 | 9088190 | ||
BC172098.1 | - | 1 | 1164 |
