siRNA Details
Gene Symbol: Gm266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033248.3 | NP_001028420.2 | 1 | 508 | ||
GenBank mRNA | AK140917.1 | - | 1 | 471 | ||
AK166394.1 | - | 1 | 482 | |||
BC145736.1 | - | 1 | 508 | 40131039 | ||
BC145762.1 | - | 1 | 508 | 40131065 |
