siRNA Details
Gene Symbol: Best2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130194.1 | NP_001123666.1 | 2, 3 | 241 | ||
GenBank mRNA | AY450428.1 | - | 2, 3 | 483 | ||
BC019528.1 | - | 1, 2 | 215 | 4208860 | ||
BC031186.1 | - | 1, 2 | 202 | 4989959 | ||
BC036157.1 | - | 2, 3 | 241 | 4982413 | ||
BC036163.1 | - | 2, 3 | 282 | 4984410 | ||
BY096795.1 | - | 2, 3 | 231 |
