siRNA Details
Gene Symbol: Ifitm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033632.1 | NP_001028804.1 | 1 | 113 | ||
GenBank mRNA | AK039626.1 | - | 1 | 129 | ||
BC104338.1 | - | 1 | 113 | 40049250 | ||
BC104339.1 | - | 1 | 113 | 40049257 |
