siRNA Details
Gene Symbol: Ifitm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033632.1 | NP_001028804.1 | 2 | 329 | ||
GenBank mRNA | AK039626.1 | - | 2 | 345 | ||
AY247203.1 | - | 1 | 59 | |||
BC104338.1 | - | 2 | 329 | 40049250 | ||
BC104339.1 | - | 2 | 329 | 40049257 |
