siRNA Details
Gene Symbol: Lemd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033250.4 | NP_001028422.2 | 2 | 107 | ||
GenBank mRNA | AK016010.1 | - | 2 | 112 | ||
AK144269.1 | - | 114 | ||||
BC048759.1 | - | 2 | 71 | |||
BC115462.1 | - | 2 | 88 | 40100287 | ||
BC115463.1 | - | 2 | 88 | 40100292 |
