siRNA Details
Gene Symbol: Szrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025608.2 | NP_001020779.1 | 4 | 498 | ||
NM_001277195.1 | NP_001264124.1 | 4 | 495 | |||
GenBank mRNA | AK027934.1 | - | 3 | 411 | ||
AK145912.1 | - | 3 | 418 | |||
AK171420.1 | - | 3 | 408 | |||
AK171702.1 | - | 3 | 423 | |||
BC066992.1 | - | 4 | 446 | 5685985 | ||
CV784607.1 | - | 4 | 437 |
Gene Symbol: 4921514A10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132662.1 | - | 1 | 115 |
