siRNA Details
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001295.2 | NP_001001295.2 | 13 | 2466 | ||
NM_001177784.1 | NP_001171255.1 | 13 | 2427 | |||
NM_172519.3 | NP_766107.1 | 13 | 2370 | |||
GenBank mRNA | AK029974.1 | - | 13 | 2371 | ||
AK032451.1 | - | 13 | 2374 | |||
AK077031.1 | - | 13 | 2430 | |||
AK150851.1 | - | 13 | 2412 | |||
AK151780.1 | - | 13 | 2412 | |||
AK153433.1 | - | 13 | 2412 | |||
AK173310.1 | - | 13 | 2400 | |||
BC056939.1 | - | 13 | 2399 | 5708685 |
