siRNA Details
Gene Symbol: Etnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175443.5 | NP_780652.2 | 6 | 1110 | ||
GenBank mRNA | AK077179.1 | - | 6 | 849 | ||
AK087818.1 | - | 5 | 908 | |||
BC152310.1 | - | 6 | 1048 | 40089976 |
