siRNA Details
Gene Symbol: Cdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145991.1 | NP_666103.1 | 10, 11 | 1021 | ||
GenBank mRNA | AK080861.1 | - | 10, 11 | 1153 | ||
BC027756.1 | - | 10, 11 | 1122 | 5067496 | ||
BC031127.1 | - | 10, 11 | 1021 | 4972826 |
