siRNA Details
Gene Symbol: Fam173a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145410.4 | NP_663385.2 | 1 | 210 | ||
GenBank mRNA | AK132031.1 | - | 1 | 84 | ||
AK146214.1 | - | 1 | 191 | |||
AK162337.1 | - | 1 | 186 | |||
BB855744.1 | - | 1 | 211 | |||
BC096050.1 | - | 1 | 105 | 5119802 |
